Learn More
CD121a Rat anti-Mouse, NA/LE, Unlabeled, Clone: 35F5, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553693
Description
The 35F5 monoclonal antibody specifically binds to the Type I Interleukin-1 Receptor (IL-1 RI) which is also known as CD121a. CD121a is expressed by T-cell, fibroblast, and epithelial cell lines such as EL-4 and 3T3. Expression of the 35F5 epitope on normal mouse thymocytes and splenocytes is undetectable by flow cytometry. CD121a is an 80 kDa type-I transmembrane glycoprotein, a member of the Ig superfamily that mediates the inflammatory responses of leucocytes to IL-1. The 35F5 antibody is capable of blocking the binding of IL-1α, IL-1β, and IL-1 receptor antagonist to the Type I IL-1 Receptor. The 35F5 mAb is highly specific for the Type I IL-1 Receptor and does not react with the Type II IL-1 Receptor (CD121b) which is present on mouse macrophage and B-cell lines and polymorphonuclear leukocytes (PMN) and PMN progenitors.

Specifications
CD121a | |
Monoclonal | |
1.0mg/mL | |
Il1r1; IL-1 Receptor Type I/p80; IL-1 R alpha; IL-1 Rα; IL-1 RI | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
35F5 | |
Unconjugated | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
IL-1 receptors from the C57BL/6 mouse T lymphoma EL4.IL2 | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.