Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD121b Rat anti-Mouse, Brilliant Violet 421, Clone: 4E2, BD

Catalog No. BDB562926 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562926 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562926 Supplier BD Biosciences Supplier No. 562926
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 4E2 monoclonal antibody specifically binds to CD121b which is also known as the IL-1 receptor type II (IL-1RII, IL-1R2) or IL-1 receptor beta (IL-1R-beta, IL-1Rβ). CD121b is encoded by the Il1r2 gene and presents as a 68 kDa type I transmembrane glycoprotein. It is expressed on a variety of cell types including B cells, neutrophils, monocytes, and bone marrow progenitor cells. A soluble form of CD121b is produced by proteolysis especially after cellular activation. Unlike the IL-1 type I receptor (CD121a), CD121b has a short cytoplasmic domain and does not mediate IL-1 signaling. CD121b may function in a membrane or soluble form as a decoy receptor that can bind IL-1 and reduce its effective concentration and biological activity. The 4E2 antibody blocks IL-1 binding to the IL-1 receptor type II.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD121b
Applications Flow Cytometry
Classification Monoclonal
Clone 4.00E+02
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Il1r2; IL-1RII; IL-1R2; Il1rb; IL-1Rβ; IL-1R-beta; IL-1 receptor beta chain
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse IL-1RII Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.