Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 Rat anti-Mouse, Alexa Fluor 647, Clone:  5H4, BD

Catalog No. BDB564924 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564924 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564924 Supplier BD Biosciences Supplier No. 564924

Rat Monoclonal Antibody

The 5H4 antibody monoclonal antibody specifically binds to mouse CD122. CD122 is a 70-85 kDa type I transmembrane glycoprotein of the hematopoietin receptor superfamily. CD122 is also known as IL-2 Receptor beta chain (IL-2R β ) or IL-15 Receptor beta chain (IL-15R β ) as it helps form signaling receptor complexes for Interleukin-2 (IL-2) or IL-15, respectively. CD122 can combine with either CD132 (γc) alone or CD132 plus CD25 (IL-2Rα) to form inte rmediate or high-affinity IL-2 Receptor complexes, respectively. The IL-2R β chain is constitutively expressed on NK cells and at lower levels on some thymocytes, T cells, B cells, monocytes, and macrophages. IL-2R β chain expression is up regulated by IL-2. The immunogen used to generate this hybridoma was rat myel oma cells transfected with a truncated IL-2R β cDNA. 5H4 does not block IL-2 binding to the IL-2 Receptor.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD122
Applications Flow Cytometry
Classification Monoclonal
Clone  5H4
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description Il2rb; Il-2Rbeta; IL-2Rβ; IL-15Rbeta; IL-2/15 Receptor-beta; IL-2/15Rbeta
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Rat
Immunogen Rat myeloma cells transfected with a truncated IL-2R β cDNA
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.