Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 Mouse anti-Human, BB515, Clone: Mik-β3, BD

Catalog No. BDB564688 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564688 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564688 Supplier BD Biosciences Supplier No. 564688
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Mik- β 3 monoclonal antibody specifically binds to CD122. CD122 is also known as IL-2/15R β because it is the shared p75 subunit (IL-12R β /IL-15R β ) of heteromeric receptors for IL-2 and IL-15. CD122 is a 70-75 kD type I transmembrane glycoprotein expressed on thymocytes, T cells, B cells, NK cells, monocytes, hematopoietic progenitor cells, fetal liver cells and stromal cells. The CD122 plays roles in the activation, differentiation and proliferation of various cell types including T cells, B cells and NK cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD122
Applications Flow Cytometry
Classification Monoclonal
Clone Mik-β3
Conjugate Brilliant Blue 515 (BB515)
Description IL2RB; IL-2Rβ; IL-2R subunit beta; IL-2RB; IL15RB; IL-2/15Rb; IL-2R p75
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human YTS Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.