Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 Rat anti-Mouse, Unlabeled, Clone: 5H4, BD

Catalog No. BDB554451 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554451 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554451 Supplier BD Biosciences Supplier No. 554451

Rat Monoclonal Antibody

The 5H4 antibody reacts with mouse CD122, the 75-85kD β chain of the mouse IL-2 receptor, but does not block IL-2 binding to the IL-2 receptor. The IL-2R β chain is constitutively expressed on a subset of CD8+ spleen T cells, on NK cells, and at lower levels on a small population of resting B cells. IL-2R β chain expression is up-regulated by IL-2. The immunogen used to generate this hybridoma was rat myeloma cells transfected with a truncated IL-2R β cDNA.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD122
Applications Flow Cytometry
Classification Monoclonal
Clone 5H4
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IL-2 and IL-15 Receptor β chain, P75
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Rat myeloma cells transfected with a truncated IL-2R β cDNA
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.