Learn More
CD122 Rat anti-Mouse, FITC, Clone: TM-β 1, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 561693
Description
The TM-β1 antibody reacts with the 90-100-kDa β chain shared by the IL-2 and IL-15 receptors (IL-2Rβ, CD122). In the periphery, CD122 is expressed on CD8+ T lymphocytes, NK cells, NK-T cells, dendritic epidermal T cells, subsets of intraepithelial lymphocytes, and macrophages. Small subsets of fetal and adult thymocytes constitutively express CD122. CD122+ cells in the bone marrow include committed NK-cell progenitors. IL-2Rβ expression is upregulated by IL-2. CD122 is a transmembrane glycoprotein of the hematopoietin receptor superfamily which can combine with CD132 (γc) alone or CD132 plus CD25 (IL-2Rα) to form intermediate or high-affinity IL-2 receptor complexes, respectively. The β chain of these complexes, CD122, is involved in signal transduction and immunoregulation. The TM- β 1 antibody blocks high affinity binding of IL-2 or IL-15 to IL-2R β .
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD122 | |
Monoclonal | |
0.5mg/mL | |
Il2rb; Il-2Rbeta; IL-2Rβ; IL-15Rbeta; IL-2/15 Receptor-beta; IL-2/15Rbeta | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
TM-β 1 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-2Rβ Transfected Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.