Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 Mouse anti-Human, PE, Clone: MIK- Tests2, BD

Catalog No. BDB554522 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554522 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554522 Supplier BD Biosciences Supplier No. 554522
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody from this clone reacts with the 75 kD β subunit (p75) of the IL-2 receptor (IL-2Rβ). Cell surface IL-2Rα molecules are expressed by T cells, B cells, monocytes, myeloid precursors, NK cells, and LGL. Together with the α subunit (p55, CD25) and the common γ subunit (γc subunit of the IL-2R, IL-4R, and IL-7R), the IL-2Rβ

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD122
Applications Flow Cytometry
Classification Monoclonal
Clone MIK- Tests2
Concentration 0.2mg/mL
Conjugate PE
Description IL-2 Receptor β chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.