Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD123 Rat anti-Mouse, Biotin, Clone: 5B11, BD

Catalog No. BDB555070 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555070 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555070 Supplier BD Biosciences Supplier No. 555070
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 5B11 monoclonal antibody specifically recognizes mouse CD123, the α chain subunit (IL-3R α ) of the mouse IL-3 receptor. The IL-3R α chain is a 60-70 kD transmembrane glycoprotein that binds IL-3 with low affinity but cannot transduce signals by itself. The mouse IL-3R α chain can complex with either of two homologous β chain subunits ( β c and β α subunits of the mouse IL-3R, IL-5R and GM-CSFR to form high-affinity receptors. The β IL-3 subunit can form high affinity IL-3 receptor complexes with only the IL-3R α subunit. The β IL-3 subunit clone is specific for mouse IL-3 and binds with low affinity. The immunogen used to generate the 5B11 hybridoma was CTLL cells transfected with the mouse IL-3R alpha chain. The 5B11 antibody does not block the binding of IL-3 protein to the high affinity IL-3 receptor heterodimer.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD123
Applications Flow Cytometry
Classification Monoclonal
Clone 5B11
Concentration 0.5mg/mL
Conjugate Biotin
Description Il3ra; IL-3RA; IL-3R-alpha; IL-3 Receptor alpha; IL-3 Receptor α; SUT-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse IL-3R transfected CTLL cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.