Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD123 Mouse anti-Human, PE, Clone: 9F5, BD

Catalog No. BDB555644 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555644 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555644 Supplier BD Biosciences Supplier No. 555644
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 9F5 monoclonal antibody specifically binds to CD123. CD123 is the 70 kDa IL-3 receptor α chain (IL-3Rα) that associates with the 120-140 kDa β subunit (CD131/Common β-chain/βc) to form the functional IL-3 receptor complex. The βc chain is also shared with distinct α chain subunits to form the functional heterodimeric receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on a subset of peripheral blood dendritic cells, myeloid precursors, basophils, mast cells, macrophages, and megakaryocytes. Reports indicate that IL-3Rα is also expressed on lymphocytes. The IL-3R plays an important role in hematopoietic progenitor cell growth and differentiation. This antibody does not block binding of IL-3 to the IL-3 receptor.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD123
Applications Flow Cytometry
Classification Monoclonal
Clone 9F5
Concentration 0.2mg/mL
Conjugate PE
Description IL-3 Receptor α chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IL-3Rα Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.