Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD124 Rat anti-Mouse, Unlabeled, Clone: MIL4R-M2, BD

Catalog No. BDB552952 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552952 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552952 Supplier BD Biosciences Supplier No. 552952

Rat Monoclonal Antibody

The mIL4R-M2 monoclonal antibody specifically binds to the α subunit of the mouse Interleukin-4 Receptor (IL-4R α , aka, CD124). The mIL4R-M2 antibody does not recognize human or rat IL-4R; crossreactivities with IL-4R from other species have not been established. The mouse IL-4R α is a 140 kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and nonhematopoietic cells and cell lines. The cell surface IL-4R α chain binds IL-4 with high affinity and associates with either the common γchain (IL-4R α / γα (sIL-4R) that result either from enzymatic cleavage of the cell surface extracellular IL-4R α domain or from differential mRNA splicing and secretion by cells. These sIL-4R retain their high-affinity ligand binding domain and appear to either enhance or inhibit IL-4-mediated functions depending on the relative local levels of IL-4 and sIL-4R. The immunogen used to generate the mIL4R-M2 hybridoma was CTLL-19.4 cells, a subclone of CTLL-2 cells that express high levels of IL-4R α .

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD124
Applications ELISA
Classification Monoclonal
Clone MIL4R-M2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IL-4 Receptor α chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen CTLL-19.4 cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.