Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D124 Rat anti-Mouse, PE, Clone: mIL4R-M1, BD

Catalog No. BDB561695 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561695 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561695 Supplier BD Biosciences Supplier No. 561695
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The mIL4R-M1 monoclonal antibody specifically binds to CD124 which is also known as the α subunit of the mouse Interleukin-4 Receptor (IL-4Rα). The mouse IL-4Rα is a 140 kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and nonhematopoietic cells and cell lines. The cell surface IL-4Rα chain binds IL-4 with high affinity and associates with either the common γ chain (IL-4Rα/γα (sIL-4R) that result either from enzymatic cleavage of the cell surface extracellular IL-4Rα domain or from differential mRNAsplicing and secretion by cells. These sIL-4R retain their high-affinity ligand binding domain and appear to either enhance or inhibit IL-4-mediated functions depending on the relative local levels of IL-4 and sIL-4R.

TRUSTED_SUSTAINABILITY

Specifications

Antigen D124
Applications Flow Cytometry
Classification Monoclonal
Clone mIL4R-M1
Concentration 0.2mg/mL
Conjugate PE
Description Il4r; IL-4Rα; Il4ra; IL-4RA; IL-4R-alpha; IL4-BP; IL-4 Receptor α chain
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CTLL-19.4 T Cell Line
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.