Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD125 Rat anti-Mouse, Alexa Fluor 488, Clone: T21, BD

Catalog No. BDB558533 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558533 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558533 Supplier BD Biosciences Supplier No. 558533
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The antibody clone T21 recognizes the mouse α-subunit (also known as CD125) of the IL-5 receptor. The IL-5 receptor is comprised of α- and β-subunits where the α-subunit is specific to the binding of IL-5 ligand and the β-subunit is common among the IL-3, IL5 and granulocyte/macrophage colony-stimulating factor (GM-CSF) receptors. Murine IL-5 belongs to the hematopoietic growth factor family of cytokines and is known to enhance several functions of murine B-cells, including immunoglobulin production, growth and differentiation. Upon binding, T21 inhibits IL-5 induced B-cell proliferation and Ig-secretion.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD125
Applications Flow Cytometry
Classification Monoclonal
Clone T21
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Description IL-5 Receptor α-subunit
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse T88-M
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.