Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD125 Mouse anti-Human, PE, Clone: A14, BD

Catalog No. BDB555902 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555902 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555902 Supplier BD Biosciences Supplier No. 555902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the α chain of the interleukin-5 receptor (IL-5R), a 55-60 kDa polypeptide, expressed on eosinophils and basophils. The IL-5R is composed of a specific α chain and a β chain which is homologous to the β chains of interleukin-3 (IL-3R) and GM-CSF receptors. IL-5 promotes the growth and differentiation of eosinophil precursors and activates mature eosinophils. Clone A14 was developed using IL-5R α -transfected cells as immunogen. This antibody binding can be blocked by IL-5.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD125
Applications Flow Cytometry
Classification Monoclonal
Clone A14
Concentration 0.2mg/mL
Conjugate PE
Description IL-5Rα
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen IL-5R α-transfected cells
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.