Learn More
CD126 Mouse anti-Human, Biotin, Clone: M182, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 552503
Description
The M182 antibody reacts with the a subunit of the human IL-6 Receptor (IL-6R α , aka, CD126). The human IL-6R α is an 80 kDa type I transmembrane glycoprotein, also known as B cell stimulatory factor-2 (BSF-2) receptor and IL-6 receptor. The IL-6R α subunit associates with the 130-160 kDa gp130 subunit (IL-6 receptor ß chain, CD130), that is shared with the receptors for Leukemia Inhibitory Factor (LIF), Ciliary Neurotropic Factor (CNTF), Oncostatin M (OSM), IL-11, Cardiotropin 1 (CT-1) and possibly Neurotrophin-1/B Cell-Stimulating Factor 3 (NNT-1/BSF-3). The IL-6R α chain binds IL-6 with low affinity, however the association with CD130 stabilizes the IL-6/IL-6R α complex resulting in the formation of a high affinity complex. The IL-6R ß chain mediates signal transduction. IL-6R α s are expressed at high levels by activated and EBV-transformed B cells, plasma cells and myeloma cells and at lower levels by most leucocytes, epithelial cells, fibroblasts, hepatocytes and neural cells. IL-6R α exists in soluble form in human serum. The serum levels of soluble IL-6Raappear to elevate in pathological situations such as multiple myeloma, Grave's disease, juvenile chronic arthritis and HIV.10-13 The immunogen used to generate the M182 hybridoma was a mixture of U266, XG-1 (human myeloma cell line expressing membrane IL-6R) and BWD41 cells (murine thymoma cell line transfected with cDNA encoding the extracellular part of IL-6R α ).

Specifications
CD126 | |
Monoclonal | |
0.5mg/mL | |
IL-6 Receptor α Chain | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
ELISA | |
M182 | |
Biotin | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mixture of U266, XG-1and BWD41 cells | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.