Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD126 Mouse anti-Human, Clone: B-R6, MilliporeSigma™

Catalog No. CBL714MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL714MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL714MI Supplier MilliporeSigma Supplier No. CBL714
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-CD126 Antibody, clone B-R6 is validated for use in ELISA, flow cytometry, immunoprecipitation, immunohistochemistry for the detection of CD126.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD126
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone B-R6
Conjugate Unconjugated
Formulation The monoclonal was purified by ion exchange chromatography and is presented as a liquid in phosphate buffered saline, pH 7.4, containing 0.09% sodium azide.
Gene Accession No. NM_000565.2; NM_181359.1
Gene Symbols IL6R, MGC104991, IL-6R-alpha, IL6RA, CD126, gp80, IL-6R-1
Host Species Mouse
Immunogen CHO cells transfected with a cDNA of the human IL-6 receptor.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.