Learn More
CD127 Rat anti-Mouse, APC, Clone: SB/199, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564175
Description
The SB/199 monoclonal antibody specifically binds to mouse CD127, the 65-75 kDa type-I transmembrane protein IL-7R α . The high affinity IL-7 receptor complex is composed of at least two transmembrane proteins, IL-7R α and CD132, the common γchain. CD127 has some sequence homology to the cytokine receptor superfamily (also known as the hematopoietin receptor superfamily). Mice lacking CD127 display profoundly impaired development of the B and T lymphoid cell lineages, but display no obvious non-lymphoid abnormalities. IL-7R α is expressed on common lymphoid progenitors and early stages of B lineage development in the bone marrow, on the earliest thymocyte progenitors, on CD4-CD8- double-negative and CD4+ and CD8+ single-positive thymocytes, and on most peripheral T lymphocytes. Intestinal intraepithelial lymphocytes with low-density γδ TCR upregulate CD127 expression in response to IL-2, which may be secreted by neighboring αβ TCR-bearing T cells.

Specifications
CD127 | |
Monoclonal | |
0.2mg/mL | |
Interleukin-7 receptor alpha chain; IL-7R alpha; IL-7RA; IL-7Rα; Il7r | |
BALB/c mouse pre-B cell line 1A9 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
SB/199 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.