Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD127 Rat anti-Mouse, Biotin, Clone: B12-1, BD

Catalog No. BDB555288 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555288 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555288 Supplier BD Biosciences Supplier No. 555288
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The B12-1 monoclonal antibody specifically binds to mouse CD127 which is also known as, the Interleukin-7 Receptor alpha chain ( IL-7Rα ). CD127 associates with CD132 ( common γchain ) to form a high-affinity signaling IL-7R complex that mediates the biological effects of IL-7. CD127 can also complex with the Thymic Stromal Lymphopoietin Receptor (TSLPR) subunit to bind and mediate cellular responses to TSLP. CD127 is a 65-75 kDa type-I transmembrane protein that belongs to the hematopoietin receptor and the Ig gene superfamilies. Mice lacking CD127 display profoundly impaired development of the B and T lymphoid cell lineages, but display no obvious non lymphoid abnormalities. IL-7Rα is expressed on common lymphoi with low-density γδ TCR expression upregulate CD127 expression in response to IL-2, which may be secreted by ne ighboring αβ TCR+ T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD127
Applications Flow Cytometry
Classification Monoclonal
Clone B12-1
Concentration 0.5mg/mL
Conjugate Biotin
Description Interleukin-7 receptor alpha chain; IL-7R alpha; IL7RA;IL-7RA; IL-7Rα; Il7r
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CD127-human IgG fusion protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a λ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.