Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD127 (IL-7Ra) Mouse anti-Human, APC, Clone: R34-34, MilliporeSigma™

Catalog No. MABF385MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF385MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF385MI Supplier MilliporeSigma Supplier No. MABF385

Mouse Monoclonal Antibody

This Mouse Monoclonal Antibody Anti-CD127 (IL-7Ra) (human), APC, clone R34-34, Cat. No. MABF385 is validated for use in FC for the detection of CD127 (IL-7Ra).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD127 (IL-7Ra)
Applications Flow Cytometry
Classification Monoclonal
Clone R34-34
Concentration 5μL (0.5μg)/test
Conjugate APC
Formulation Purified Mouse Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols IL7R
Host Species Mouse
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.