Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD127 (IL-7Ra) Rat anti-Mouse, FITC, Clone: A7R34, MilliporeSigma™

Catalog No. MABF380MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF380MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF380MI Supplier MilliporeSigma Supplier No. MABF380
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Rat Monoclonal Antibody Anti-CD127 (IL-7Ra) (mouse), FITC, clone A7R34, Cat. No. MABF380 is validated for use in immunoprecipitation, immunohistochemistry, immunofluoresence, FC for the detection of CD127 (IL-7Ra).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD127 (IL-7Ra)
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone A7R34
Concentration 0.5mg/mL
Conjugate FITC
Formulation Purified Rat Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols Il17ra, Il17r
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 1 Unit
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.