Learn More
CD13 Rat anti-Mouse, FITC, Clone: R3-242, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 558744
Description
The R3-242 monoclonal antibody specifically binds to aminopeptidase N (APN), which has been reported to be the mouse CD13 homologue. APN is expressed in a variety of mammalian tissues, including intestinal and renal epithelial brush border, endothelium, liver, lung, and brain. In lymphoid tissues, R3-242 mAb is reported to detect APN on monocytes, macrophages, mast cells, dendritic cells, and B lymphocytes, but not on T cells, and thymocytes. However, the reported reactivity with B cells may be non-specific, via binding to the Fcγ receptors (CD16/CD32). This observation is in agreement with reports using different antibodies to mouse APN. APN may play a role in antigen presentation in the immune system. R3-242 mAb does not inhibit the enzymatic activity of APN.
Specifications
| CD13 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Crude Aminopeptidase N Preparation from BALB/c Mouse Intestinal Epithelium | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| R3-242 | |
| FITC | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.