Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD131 Mouse anti-Human, PE, Clone: 3D7, BD

Catalog No. BDB564190 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564190 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564190 Supplier BD Biosciences Supplier No. 564190

Mouse Monoclonal Antibody

The 3D7 antibody reacts with CD131, the 120 kD common β chain (βc) which is shared with the receptor complexes for human granulocyte-macrophage colony stimulating factor (GM-CSFR), interleukin-3 (IL-3R) and interleukin-5 (IL-5R). Together with the α subunit of either the IL-3R (IL-3Rα), IL-5R (IL-5Rα), or GM- CSFR (GM-CSFRα), the common ββ chains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD131
Applications Flow Cytometry
Classification Monoclonal
Clone 3D7
Conjugate PE
Description Cytokine receptor common β chain; CDw131; IL-3Rβ; IL-5Rβ
Host Species Mouse
Immunogen Human CD131 Transfected COS Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.