Learn More
D131 Rat anti-Mouse, PE, Clone: JORO50 (also known as JORO 50), BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559920
Description
The JORO50 monoclonal antibody specifically recognizes the extracellular regions of the mouse CD131/ βc (Bc, AIC2B, Il3rb1, Csf2rb1) and βIL3 (BIL3, AIC2A, Il3rb2, Csf2rb2) cytokine receptor subunits. A variety of mouse cell ty pes, including multipotential hematopoietic stem cells, mast cells, megakaryocytes, eosinophils, erythroblasts, pre-B cells, and osteoclasts, express βIL-3 and βc subunits. Either βIL-3 or βc can combine with the IL-3Rα chain to form two distinct, functional (i.e., signaling), high affinity receptors for mouse IL-3. The βIL-3 subunit by itself can bind mouse IL-3 with low affinity whereas the βc subunit can not. βc (but not βIL-3) can combine with the IL-5Rα (CD125) and GM-CSFRα (CD116) subunits to form hig h affinity, signaling receptors for mouse IL-5 or GM-CSF, respectively. The immunogen used to generate the JORO50 hybridoma was the bone marrow-derived, C4-77 pro-T lymphocyte clone.
Specifications
| D131 | |
| Monoclonal | |
| 0.2mg/mL | |
| CD131, βc, Bc, AIC2B, Il3rb1, Csf2rb1; βIL3, BIL3, AIC2A, Il3rb2, Csf2rb2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| JORO50 (also known as JORO 50) | |
| PE | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Bone Marrow-Derived C4-77 Pro-T Lymphocyte Clone | |
| 0.2 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.