Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Rat anti-Mouse, NA/LE, Unlabeled, Clone: 4G3, BD

Catalog No. BDB554454 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554454 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554454 Supplier BD Biosciences Supplier No. 554454
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 4G3 antibody reacts with the common γ subunit (γc) shared by the IL-2, IL-4, IL-7, IL-9 and IL-15 receptors also known as CD132. This receptor is expressed constitutively at low levels on most lymphocytes, myeloid cells, and embryonic thymocytes. The γc receptor is a 75 - 80kDa transmembrane glycoprotein which mediates signal transducing activities of cytokine receptor complexes with which it is associated.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD132
Applications Flow Cytometry
Classification Monoclonal
Clone 4G3
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Common γ chain, γc
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.