Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Rat anti-Mouse, PE, Clone: 4G3, BD

Catalog No. BDB554457 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554457 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554457 Supplier BD Biosciences Supplier No. 554457
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 4G3 monoclonal antibody specifically binds to the common γsubunit ( γc) shared by the IL-2, IL-4, IL-7, IL-9, IL-15, and IL-21 receptors also known as CD132. This receptor is expressed constitutively at low levels on most lymphocytes, myeloid cells, embryonic thymocytes and lymphoid cell lines, but not non-lymphoid cells. The γc receptor is a 75 - 80 kD transmembrane glycoprotein which mediates signal transducing activities of cytokine receptor complexes with which it is associated.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD132
Applications Flow Cytometry
Classification Monoclonal
Clone 4G3
Concentration 0.2mg/mL
Conjugate PE
Description Il2rg; IL-2Rγ; Commin gamma chain; Common γ; gamma c receptor; γc
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.