Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Rat anti-Human, PE, Clone: TUGH4, BD

Catalog No. BDB555898 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555898 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555898 Supplier BD Biosciences Supplier No. 555898
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TUGh4 monoclonal antibody specifically binds to CD132. CD132 is a 65-70 kDa type 1 transmembrane glycoprotein that is encoded by the IL2RG (interleukin 2 receptor, gamma) gene. CD132 is also known as the common γsubunit (γc) and is shared by the IL-2, IL-4, IL-7, IL-9, IL-15 and IL-21 receptor complexes. CD132 is broadly expressed by most peripheral T and B lymphocytes, NK cells, monocytes, and granulocytes. The cytoplasmic domain of the γc chain plays an important role in cytokine-mediated signal transduction. Mutation of the IL2RG gene results in X-linked severe combined immunodeficiency (XSCID). The TUGh4 antibody recognizes a different epitope from that recognized by the CD132-specific clone, AG184.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD132
Applications Flow Cytometry
Classification Monoclonal
Clone TUGH4
Concentration 0.2mg/mL
Conjugate PE
Description IL-2RG; IL-2Rγ; γc; Common γ chain; Common gamma chain; gamma c; SCIDX1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Human γc Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.