Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD133 Rat anti-Mouse, AlexaFluor™488, Clone: 13A4, MilliporeSigma™

Catalog No. MAB4310XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4310XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4310XMI Supplier MilliporeSigma Supplier No. MAB4310X
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-CD133 Antibody, clone 13A4, Alexa Fluor 488 conjugated is an antibody against CD133 for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD133
Applications Flow Cytometry
Classification Monoclonal
Clone 13A4
Conjugate Alexa Fluor 488
Formulation Liquid in Phosphate buffer with 15mg/mL BSA as a stabilizer and 0.1% sodium azide.
Gene Accession No. NM_006017.1
Gene Symbols PROM1, CD133, AC133, MSTP061, hProminin, PROML1
Host Species Rat
Immunogen Telencephali of ten 12-day-old NMRI mouse embryos (E12, ∼1 mg protein), which consist mostly of neuroepithelium, were homogenized in 1mL PBS, mixed with crushed nitrocellulose filter (1 cm2 in 0.5mL PBS) as adjuvant, and injected i.p. into a
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 24 months. Protect from Light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.