Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD134 Mouse anti-Human, Brilliant Violet 711, Clone: ACT35 (also known as Ber-ACT35), BD

Catalog No. BDB563664 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563664 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563664 Supplier BD Biosciences Supplier No. 563664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ACT35 monoclonal antibody specifically binds to CD134 which is also known as OX40. The 35 kDa CD134 polypeptide is encoded by the TNFRSF4 gene. CD134 is a type I integral membrane glycoprotein and member of the tumor necrosis factor/nerve growth factor receptor (TNFR/NGFR) family. CD134 is expressed on activated T lymphocytes, hematopoietic precursor cells and fibroblasts. CD134 functions as a T cell costimulatory receptor when bound by OX40 Ligand/TNFSF4 that is expressed by antigen presenting cells. CD134 thereby plays roles in T-cell activation as well as the regulation of differentiation, proliferation or apoptosis of normal and malignant lymphoid cells. Analysis of the nucleotide sequence of the human TNFRSF4 cDNA reveals strong homology with the rat Tnfrsf4 cDNA sequence. OX40 was clustered as CD134 in the Sixth International Workshop on Human Leukocyte Differentiation Antigens.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD134
Applications Flow Cytometry
Classification Monoclonal
Clone ACT35 (also known as Ber-ACT35)
Conjugate Brilliant Violet 711
Description OX40; TNFRSF4; TXGP1L; OX40L Receptor
Host Species Mouse
Immunogen Human HUT 102
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.