Learn More
CD134 Mouse anti-Human, PE, Clone: L106, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340420
Description
CD134, clone L106, is derived from hybridization of mouse Sp2/0 cells with spleen cells from BALB/c mice immunized with phytohemagglutinin-activated T lymphocytes.
CD134 (Anti-Human Ox 40) recognizes a 50-kdalton (kDa) antigen that is present on activated T lymphocytes. By sequence analysis, it is related to the rat monoclonal antibody Ox 40, a member of the nerve growth factor superfamily that also includes CD27, CD30, CD40, CD95 (Fas), and the tumor necrosis factors alpha and beta (CD120a and CD120b).

Specifications
CD134 | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
L106 | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Phytohemagglutinin-activated T lymphocytes | |
50 Tests | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.