Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD135 Rat anti-Mouse, Brilliant Violet 421, Clone: A2F10.1, BD

Catalog No. BDB562898 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562898 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562898 Supplier BD Biosciences Supplier No. 562898
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The A2F10 monoclonal antibody specifically binds to Flk-2/Flt3 (Ly-72, CD135), a receptor protein tyrosine kinase closely related to c-kit, c-fms, and PDGF Receptor of the immunoglobulin superfamily. The Flt3 message is detected in hematopoietic stem cells and primitive progenitor cells in fetal liver, adult bone marrow, and fetal and adult thymus, as well as brain, placenta, and testis; but it is absent in more mature hematopoietic cells. In flow cytometric analysis, the A2F10 antibody recognizes Flt3 -transfected Y3 cells (rat myeloma), but not the parent cell line in addition to recognizing early B lymphoid lineage cells in juvenile and adult bone marrow. A role for CD135 in the regulation of hematopoiesis is suggested by the observations that soluble Flk-2/Flt3 ligand can both stimulate proliferation of stem cell-enriched fetal liver, fetal thymus, and adult bone marrow populations and enhance their responses to other growth factors in vitro . In addition, injection of Flk-2/Flt3 ligand stimulates extramedullary hematopoiesis in the mouse spleen and accumulation of dendritic cells in the hematopoietic system. mAb A2F10.1 is reported to immunoprecipitate a 150-kDa surface protein from the murine myeloblast cell line M1, which naturally expresses CD135, and to inhibit the binding of Flk-2/Flt3 ligand to CD135.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD135
Applications Flow Cytometry
Classification Monoclonal
Clone A2F10.1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Flt3; Fms-like tyrosine kinase 3; FLT-3; FLK-2; Fetal liver kinase 2; Ly72
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Flt-3 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.