Learn More
Description
The 4B4-1 monoclonal antibody specifically binds to CD137 which is also known as 4-1BB, and ILA (induced by lymphocyte activation). CD137 is a type I transmembrane glycoprotein that belongs to the TNF/NGF receptor family. It is encoded by TNFRSF9 (tumor necrosis factor receptor superfamily, member 9). CD137 is expressed on activated T cells, B cells, monocytes, and follicular dendritic cells. CD137 plays roles in the costimulation, differentiation and survival of T cells and B cells.
Specifications
Specifications
| Antigen | CD137 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4B4-1 |
| Conjugate | Brilliant Violet 421 |
| Description | 4-1BB |
| Host Species | Mouse |
| Immunogen | Recombinant Human 4-1BB Fusion Protein |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.