Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Ligand Mouse anti-Human, Unlabeled, Clone: C65-485, BD

Catalog No. BDB559445 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559445 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559445 Supplier BD Biosciences Supplier No. 559445
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a type II membrane protein known as 4-1BB ligand. This molecule belongs to the TNF superfamily. It has been reported to have costimulatory function on activated T cells. Reports suggest that stimulation of activated T cells via 4-1BB ligand does not require the presence of other costimulatory molecules, including CD28. Some studies have reported human 4-1BB ligand to be expressed preferentially on primary B cells and B-cell lines. C65-485 antibody is able to react with recombinant human 4-1BB ligand (rh4-1BBL) bound to CD137 (4-1BB receptor) expressed on PHA-stimulated PBMC.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD137 Ligand
Applications Flow Cytometry
Classification Monoclonal
Clone C65-485
Concentration 0.5mg/mL
Conjugate Unconjugated
Description 4-1BB Ligand
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.