Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD138 Mouse anti-Human, APC, Clone: MI15, BD

Catalog No. BDB347193 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347193 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347193 Supplier BD Biosciences Supplier No. 347193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD138 clone MI15 is derived from the fusion of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with human cell line 8266.

The CD138 antibody recognizes syndecan-1, a member of the syndecan family of transmembrane heparin sulfate proteoglycans whose main functions are to modulate ligand-dependent activation of primary signaling receptors at the cell surface. Syndecan-1 exhibits molecular polymorphism; its core protein is a single-chain molecule of 70 to 75 kilodaltons (kDa), and its isoforms vary in molecular weight up to approximately 400 kDa.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD138
Applications Flow Cytometry
Classification Monoclonal
Clone MI15
Concentration 25.0μg/mL
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human U266 and XG-1 Myeloma Cell Lines
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.