Learn More
CD138 Mouse anti-Human, Unlabeled, Clone: DL-101, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550804
Description
Reacts with CD138 (Syndecan-1), an 85-92 kDa single chain transmembrane protein, expressed on multiple myeloma-derived cell lines and malignant plasma cell populations. It is also expressed on pre-B cells, immature B cells and plasma cells, but not on mature circulating B-lymphocytes. Syndecan-1 is a member of the family of transmembrane heparan sulfate proteoglycans. It is also expressed on some non-hematopoietic cells, including embryonic mesenchymal cells, vascular smooth muscle cells, endothelial and neural cells. CD138 binds to many extracellular matrix proteins through its heparan sulfate side-chains, like fibronectin, collagen types I, III and V, tenascin, thrombospondin, and antithrombin III. It is considered an extracellular matrix receptor that may serve as a co-receptor for fibroblast growth factor and related molecules. Monoclonal antibody DL-101 does not block the binding of clone B-B4 (another anti-syndecan-1 antibody) or Mi15 by flow cytometric analysis.

Specifications
CD138 | |
Monoclonal | |
0.5mg/mL | |
Syndecan-1 | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
DL-101 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.