Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD138; Clone: MI15; BD
Supplier: BD Biosciences 563294
Description
Host Species: Mouse
Clone MI15
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Human U266 and XG-1 Myeloma Cell Lines
Flow Cytometry

Specifications
CD138 | |
MI15 | |
Human U266 and XG-1 Myeloma Cell Lines | |
50 Tests | |
Primary | |
IgG1 |
Monoclonal | |
Mouse | |
Affinity Purified | |
RUO | |
Human |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction