Learn More
Description
Host Species: Mouse
Clone: SB55a
Isotype: IgG2a
Cross Reactivity: Human
Immunogen: Recombinant human CD138
Flow Cytometry, Immunohistochemistry
Specifications
Specifications
| Antigen | CD138 |
| Classification | Monoclonal |
| Clone | SB55a |
| Conjugate | FITC |
| Host Species | Mouse |
| Immunogen | Recombinant human CD138 |
| Purification Method | Unpurified |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
