Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Mouse anti-Human, APC-H7, Clone: MΦP9, BD

Catalog No. BDB641394 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB641394 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB641394 Supplier BD Biosciences Supplier No. 641394
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD14 clone MφP9 is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with peripheral blood monocytes from a patient with rheumatoid arthritis.

The CD14 antibody recognizes a human monocyte/macrophage antigen of 55 kilodaltons (kDa).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD14
Applications Flow Cytometry
Classification Monoclonal
Clone MΦP9
Concentration 25μg/mL
Conjugate APC-H7
Formulation Phosphate buffered saline with BSA and ProClin™.
Host Species Mouse
Immunogen Human Monocytes
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.