Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Rat anti-Mouse, NA/LE, Unlabeled, Clone: 4C1/CD14, BD

Catalog No. BDB557896 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557896 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557896 Supplier BD Biosciences Supplier No. 557896
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 4C1/CD14 antibody reacts with CD14, a ∽55-kDa glycosyl phosphatidyl inositol (GPI)-linked cell-surface glycoprotein. Flow cytometric analysis demonstrates that 4C1/CD14 antibody stains CD14 transfectants, the monocyte/macrophage cell line RAW 264.7 peritoneal exudate neutrophils and macrophages, and cultured bone marrow-derived macrophages, but not undifferentiated bone marrow cells, splenocytes, or peripheral blood leukocytes. This staining pattern is similar to that of the alternate anti-mouse CD14 mAb rmC5-3, which recognizes a different CD14 epitope, and differs from that of the human, where CD14 expression is characteristic of circulating monocytes and neutrophils. Therefore, data suggest that CD14 expression by leukocyte populations may differ in mice and humans. CD14 is a receptor for the complex of lipopolysaccharide (LPS or endotoxin, from gram-negative bacteria) with LPS-binding protein (LBP, a plasma protein). After binding to LPS-LPB, CD14 associates with integral membrane proteins (such as TLR4-MD-2) which mediate signal transduction. It is involved in the development of endotoxic shock, innate immunity, inflammatory responses, and LPS-stimulated bone resorption. Unlike mAb rmC5-3, the 4C1/CD14 antibody blocks the binding of LPS to CD14.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD14
Applications Flow Cytometry
Classification Monoclonal
Clone 4C1/CD14
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen Mouse monocyte/macrophage cell line
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.