Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Mouse anti-Human, Unlabeled, Clone: M5E2, BD

Catalog No. BDB550376 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550376 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550376 Supplier BD Biosciences Supplier No. 550376
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD14
Applications Flow Cytometry
Classification Monoclonal
Clone M5E2
Concentration 250μg/mL
Conjugate Unconjugated
Description LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD14 Protein
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.