Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD140b (pY1009) Mouse, Alexa Fluor 647, Clone: J25-650, BD

Catalog No. BDB558323 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558323 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558323 Supplier BD Biosciences Supplier No. 558323
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Platelet-derived growth factor (PDGF) is a potent mitogen for cells of mesenchymal origin and exerts its effects by binding to the PDGF receptor (PDGFR), a transmembrane protein tyrosine kinase. PDGFR is composed of PDGFRα (CD140a) and/or PDGFRβ (CD140b) polypeptides. Both PDGF and PDGFR consist of subunits that form homo- or heterodimers with varying specificities: PDGF-AA binds only to αα PDGFR, PDGF-AB binds to both αα and αβ PDGFR, and PDGF-BB binds to all three PDGFRs. Ligand binding induces dime rization and activation of the receptor. Upon activation, CD140b is phosphorylated at multiple tyrosine sites and, in turn, an intracellular phosphorylation cascade is initiated. PDGFR localizes primarily to membrane invaginations termed caveolae, compar tments that are enriched in several of its downstream effectors, including phosphatidylinositol 3'-kinase, Src, and phospholipase C-γ(PLC-γ).

The J25-602 monoclonal antibody recognizes the phosphorylated tyrosine 1009 (pY1009) in the C-terminal noncataly tic region of CD140b, which interacts primarily with protein tyrosine phosphatase 1D and possibly with PLC-γ. The orthologous phosphorylation site in mouse PDGFRβ is Y1008.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD140b (pY1009)
Applications Flow Cytometry
Classification Monoclonal
Clone J25-650
Conjugate Alexa Fluor 647
Description CD140b (pY1009)
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human PDGFRβ (pY1009) Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.