Learn More
Description
Host Species: Mouse
Clone 1A4
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Purified Human Thrombomodulin
Flow Cytometry
Specifications
Specifications
| Antigen | CD141 |
| Classification | Monoclonal |
| Clone | 1A4 |
| Conjugate | Brilliant Violet 510 |
| Host Species | Mouse |
| Immunogen | Purified Human Thrombomodulin |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.