Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD144 Mouse anti-Human, Alexa Fluor 647, Clone: 55-7H1, BD

Catalog No. BDB561567 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561567 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561567 Supplier BD Biosciences Supplier No. 561567
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 55-7H1 antibody reacts with a calcium-independent epitope on cadherin-5, a member of the cadherin family of calcium-dependent adhesion molecules. Cadherin-5 is expressed on endothelial cells in vivo and in vitro. It may play a role in the organization of lateral endothelial junctions and in the control of permeability properties of vascular endothelium.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD144
Applications Flow Cytometry
Classification Monoclonal
Clone 55-7H1
Conjugate Alexa Fluor 647
Description VE-cadherin; Cadherin-5; CDH5; Vascular endothelial cadherin
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.