Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD146 Mouse anti-Human, FITC, Clone: P1H12, BD

Catalog No. BDB560846 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560846 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560846 Supplier BD Biosciences Supplier No. 560846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The P1H12 antibody reacts with a 118 kDa transmembrane glycoprotein also known as MCAM, MUC18, or Mel-CAM. CD146 is a member of the immunoglobulin superfamily and is expressed by angioblasts and mesenchymal stems cells and is strongly expressed by blood vessel endothelium and smooth muscle. CD146 is also expressed by melanoma cells, intermediate trophoblasts and a subpopulation of activated T cells. The P1H12 monoclonal antibody has been reported to block endothelial cell adhesion that is observed very early in embryogenesis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD146
Applications Flow Cytometry
Classification Monoclonal
Clone P1H12
Conjugate FITC
Description MCAM; MELCAM; MUC18; Gicerin; Melanoma cell adhesion molecule
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Umbilical Vein Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.