Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD147 Mouse anti-Human, FITC, Clone: HIM6, BD

Catalog No. BDB555962 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555962 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555962 Supplier BD Biosciences Supplier No. 555962
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIM6 monoclonal antibody specifically binds to CD147 which is encoded by BSG. CD147 is a type I transmembrane glycoprotein (30-50 kDa) of the immunoglobulin super-gene family. Neurothelin, a blood-brain barrier-specific molecule, was clustered as CD147 in the Sixth Human Leukocyte Differentiation Antigen (HLDA) workshop. It bears homology with mouse gp42 or basigin, human “M6” or “EMMPRIN”, rat OX-47 or CD-9, and avian HT7 or 5A11. CD147 is also known as Tumor cell-derived collagenase stimulatory factor (TCSF). CD147 is a molecule that is broadly expressed on cells of hematopoietic and non-hematopoietic origin. Its expression on specific cell types may be regulated by cytokines. CD147 plays a role in embryonal blood-brain barrier development and a role in integrin-mediated adhesion in brain endothelia.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD147
Applications Flow Cytometry
Classification Monoclonal
Clone HIM6
Conjugate FITC
Description BSG; Basigin; BASI; Neurothelin; 5F7; EMMPRIN; M6; OK; TCSF
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.