Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD148 Mouse anti-Human, Alexa Fluor 647, Clone: A3, BD

Catalog No. BDB564732 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564732 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564732 Supplier BD Biosciences Supplier No. 564732
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A3 monoclonal antibody specifically binds to CD148. CD148 is a receptor-type tyrosine phosphatase that belongs to the receptor class 3 subfamily of the protein-tyrosine phosphatase family. CD148 is also known as Density-enhanced phosphatase 1 (DEP-1), Human protein tyrosine phosphatase- η (HPTP- η /HPTP-eta), or Protein tyrosine phosphatase receptor type J (PTPRJ). CD148 is a highly glycosylated type I transmembrane protein that is widely expressed on different cell types including monocytes, granulocytes, dendritic cells, thymocytes, T cells, B cells, NK cells, fibroblasts, and platelets. As a protein tyrosine phosphatase with receptor function, CD148 plays a role in regulating the signaling activities of various phosphorylated cellular proteins including the PDGF Receptor, catenins and Src family kinases. In the immune system, CD148 plays a role in regulating the activities of different cell types. For example, upon T cell activation, CD148 expression is upregulated by T cells. CD148 can subsequently dephosphorylate LAT and PLC-γsignaling protein s and thereby downregulate T cell signaling responses. Immobilized A3 antibody can reportedly augment the proliferation of anti-CD3 antibody-stimulated peripheral blood T cells in culture.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD148
Applications Flow Cytometry
Classification Monoclonal
Clone A3
Conjugate Alexa Fluor 647
Description DEP-1; DEP1; HPTP eta; HPTP-η; PTPRJ; R-PTP-eta; R-PTP-J; SCC1; p260
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Activated human peripheral blood mononuclear cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.