Learn More
CD15 Mouse anti-Human, Unlabeled, Clone: W6D3, BD
Catalog No. BDB557895 Shop All BD Biosciences Products
Change view
Click to view available options
0.1mg; Unlabeled
Catalog No.
BDB557895 0.1mg; Unlabeled
1 options
Catalog No. BDB557895 Supplier BD Biosciences Supplier No. 557895
Add to Cart
Add to Cart
Mouse Monoclonal Antibody
The W6D3 monoclonal antibody specifically binds to 3-fucosyl-N-acetyllactosamine (3-FAL), a 220 kDa carbohydrate structure, also called X-hapten. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and on monocytes to a varying degree, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. Most CD15 antibodies are IgM isotype; clone W6D3 is a mouse IgG1 isotype. In comparison studies with clone HI98, a known CD15 antibody, clone W6D3 shows brighter fluorescence staining and its binding can be blocked by clone HI98.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY
For Research Use Only.