Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15 Mouse anti-Human, FITC, Clone: W6D3, BD

Catalog No. BDB562370 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562370 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562370 Supplier BD Biosciences Supplier No. 562370
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The W6D3 monoclonal antibody specifically binds to 3-fucosyl-N-acetyllactosamine (3-FAL), a 220 kDa carbohydrate structure, also called X-hapten. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and on monocytes to a varying degree, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. Most CD15 antibodies are IgM isotype; clone W6D3 is a mouse IgG1 isotype. In comparison studies with clone HI98, a known CD15 antibody, clone W6D3 shows brighter fluorescence staining and its binding can be blocked by clone HI98.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15
Applications Flow Cytometry
Classification Monoclonal
Clone W6D3
Conjugate FITC
Description 3-fucosyl-N-acetyllactosamine; 3-FAL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.