Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD150 Rat anti-Mouse, Brilliant Violet 421, Clone: Q38-480, BD

Catalog No. BDB562811 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562811 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562811 Supplier BD Biosciences Supplier No. 562811
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The Q38-480 monoclonal antibody specifically binds to mouse CD150, also known as SLAM (signaling lymphocyte activation molecule). CD150 is a type 1 transmembrane glycoprotein that is a member of the CD2 subfamily of the Ig superfamily. It is encoded by the Slamf1 (signaling lymphocytic activation molecule family member 1) gene. CD150 is differentially expressed on subsets of thymocytes, T and B lymphocytes, dendritic cells, macrophages, and endothelial cells. SLAM plays multiple roles in innate and adaptive immunity serving as an adhesion molecule and/or coreceptor. CD150-mediated costimulation of TCR-activated T cells reportedly results in the increased production of IFN- γby Th1 cells and is required for IL-4 production by T follicular helper cells. CD150 also plays important roles in hematopoietic cell developmental pathways. CD150 is differentially expressed by self-renewing adult hematopoietic stem cells (HSC) that are CD150+ whereas non-multipotent hematopoietic progenitor cells are CD150-. Utilizing additional cell surface markers, lineage-negative CD150+CD48-CD41- cell fractions are reported to be highly enriched for adult HSC.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD150
Applications Flow Cytometry
Classification Monoclonal
Clone Q38-480
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description SLAM; Slamf1; Signaling lymphocytic activation molecule family member 1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CD150 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.