Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD150 Mouse anti-Human, PE, Clone: A12, BD

Catalog No. BDB559592 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559592 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559592 Supplier BD Biosciences Supplier No. 559592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A12 monoclonal antibody specifically binds to CD150, a single chain type I transmembrane glycoprotein of approximately 75-95 kDa. CD150 is encoded by the SLAMF1 gene and is also known as a signaling lymphocyte activation molecule (SLAM), or IPO-3. It is expressed on T cells, B cells, thymocytes, germinal center and dendritic cells, but not on monocytes. Expression of CD150 is upregulated on T, B and dendritic cells after activation via T-cell and B-cell costimulatory molecules. It has been reported that CD150 plays a role in signal transduction.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD150
Applications Flow Cytometry
Classification Monoclonal
Clone A12
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Activated Human Peripheral Blood Mononuclear Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.