Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD151 Mouse anti-Human, Clone: TS151, MilliporeSigma™

Catalog No. MABT59MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT59MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT59MI Supplier MilliporeSigma Supplier No. MABT59

Mouse Monoclonal Antibody

Anti-human CD151 clone TS151 Antibody detects level of human CD151 clone TS151 and has been published and validated for use in FC and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD151
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone TS151
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001034579
Gene Symbols CD151, SFA1, SFA-1, TSPAN24, PETA-3, RAPH, tspan-24, Tspan-24, GP27, MER2, PETA3
Host Species Mouse
Immunogen Whole HeLa cells
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.