Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD152 Mouse anti-Human, APC, Clone: BNI3, BD

Catalog No. BDB560938 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560938 25 Tests
BDB555855 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560938 Supplier BD Biosciences Supplier No. 560938
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the “cytolytic T lymphocyte-associated antigen”, CTLA-4. CTLA-4 is transiently expressed on activated CD28+ T cells and binds to CD80 and CD86 present on antigen presenting cells (APC) with high avidity. This interaction appears to deliver a negative regulatory signal to the T cell. There are recent reports that indicate that CTLA-4 is also expressed on B cells when cultured with activated T cells, suggesting a possible role of CTLA-4 in the regulation of B-cell response. Immobilized BNI3.1 enhances T-cell proliferation induced by CD3 and CD28. Recent studies have showed that CD152 can be expressed by regulatory T (Treg) cells. It has been found this antibody can stain the
intracellular CD152 on the Treg cells after fixation and permeabilization of cells.

Host Species: Mouse
Clone: BNI3
Isotype: IgG2a κ
Species Reactivity: Human

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD152
Applications Flow Cytometry
Classification Monoclonal
Clone BNI3
Conjugate APC
Description CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CTLA4 Recombinant Protein
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.